Mixing measuring cylinders, tall form, class B, borosilicate glass, SILBERBRAND ETERNA
Supplier: Brand
33962 33908 33920 33928 33938 33948 33954
612-5044EA 99.3 EUR
612-5044 612-5038 612-5039 612-5040 612-5041 612-5042 612-5043
Mixing measuring cylinders, tall form, class B, borosilicate glass, SILBERBRAND ETERNA
Cylinders Measuring Cylinders
Borosilicate glass 3.3, class B.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5043 and 612-5044 octagonal PE stopper)
  • ETERNA amber coloured line graduations
* Height without stopper.
Certifikace: DIN EN ISO 4788