Repair set
Supplier: Brand
701964 701965 701966 701967 701968
612-5381EA 46.7 EUR
612-5381 612-5382 612-5383 612-5384 612-5385
Repair set
Tool Sets
Set includes 1 allen key, 1 piston rod with fitted PTFE Transferpettor™ seal (for capacities ≥ 20 µl), 1 calibration gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE Transferpettor™ seals, 1 mounting block (for capacities ≥ 20 µl).