Injection spikes for multidose vials, Mini Spike® 2
Supplier: B. Braun
4550590 4550591 4550592
612-5481EA 343 EUR
612-5481 612-5482 612-5483
Injection spikes for multidose vials, Mini Spike® 2
Infusions
Mini Spike 2 withdrawal and injection spikes of multidose vials for multiple withdrawals. A fluid filter retains particles for Mini-Spike 2 Chemo and PureSite build a closed system for patient and user safety.
  • Integrated air filter
  • Swabable needle-free access valve
  • Compatible with Luer and Luer-Lock connection
  • Protection against microbial and chemical contamination
  • Automatic closure during disconnection
  • Protection against sharps injury through needle-free technology
MiniSpike 2: The bacteria-proved air filter protects the drug against airMore Product Information