VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
612-5504EA 104 EUR
612-5504 612-5505 612-5506 612-5507 612-5521 612-5522 612-5523 612-5539 612-5540 612-5541 612-5542 612-5543 612-5544 612-5545 612-5546 612-5547 612-5548 612-5826 612-5827 612-5828 612-5833 612-5830 612-5834 612-5835 612-5831 612-5836 612-5829 612-5837 612-5832
VWR® Premium Line, Serological Pipettes
Pipettes Serological Pipettes
PS, transparent, graduated, non sterile or sterile. These disposable pipettes for liquid transfers are made of GPPS (general polystyrene), in compliance with USP Class VI.
  • DNase-/RNase-free, non pyrogenic, non cytotoxic, non haemolytic
  • Meet the requirement of BST/TSE; latex-free
  • Graduations are calibrated for accurate dispensing to within ±2%
  • Pipettes are colour coded according to their volume
  • Sterile pipettes: Sterilised by irradiation
  • Printed with a batch number for traceability
Manufactured in a class 100 000 cleanroom environment. ManufacturedMore Product Information