VWR® Automatic Burettes According to Dr. Schilling
Supplier: VWR Collection
612-6697EA 72 EUR
612-6697 612-6699 612-6700
VWR® Automatic Burettes According to Dr. Schilling
Burettes
Borosilicate 3.3 glass, class B.
  • Schellbach stripe for accurate meniscus reading
  • High contrast blue enamel graduations
  • Complete with 1000 ml plastic bottle, plastic clamp, plastic stand and PTFE pipe
  • Automatic zeroing, squeezing bottle allows rapid filling
Mainly used in chemical laboratories to measure small volumes of liquid in titration. The built-in overflow automatically drains excess liquid back to the reservoir bottle.
Certifikace: DIN EN ISO 385, error limits according to class B.