VWR®, Bottle Top Dispensers
Supplier: VWR Collection
612-6857EA 271 EUR
612-6857 612-6858 612-6859 612-6860 612-6861 612-6862
VWR®, Bottle Top Dispensers
Bottle Top Dispensers
Designed with ergonomics and intuitive handling in mind, VWR® Bottle Top Dispensers exhibit excellent chemical compatibility and offer reliable precision in practical laboratory environments.
  • Ergonomically designed
  • Excellent chemical compatibility; dispensers can also be used with solvents
  • Re-circulation valve to prevent loss of reagents
  • Springless Valve™ for jam-free dispensing
  • PTFE piston with ETP O-Ring
  • Volume adjustment knob enables effortless volume setting
  • Flexible delivery nozzle
  • Fully autoclavable at 121 °C and 15 psi for a duration of 10 to 15