Top tilt measures
Supplier: Lenz Laborglas GmbH & CO.KG
05476101 05476104 05476110 05476113 05476119 05476122 05476128 05476137 05476107
612-9850EA 42.3 EUR
612-9850 612-9851 612-9852 612-9853 612-9855 612-9856 612-9857 612-9858 201-1077
Top tilt measures
Bottle Top Tilt Measures
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These tilt measures with ground joint cone NS 29/32 can fit flasks - see 215-1117 and 215-1118.