Reagent reservoirs
Supplier: Thermo Fisher Scientific
95128093 95128095 95128085 95128080
613-0828EA 146 EUR
613-0828 613-0829 613-0827 613-0826
Reagent reservoirs
Reagent Reservoirs
White PS. These disposable reservoirs feature graduations on their inside wall which enable a quick measurement of the remaining liquid.
  • Trough within a trough maximises the amount of liquid accessible to pipette tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25 ml divided reservoir allows the pipetting of two different