Bottle top burettes
Supplier: VITLAB
1620506 1620507
613-2167EA 896 EUR
613-2167 613-2168
Bottle top burettes
Bottle Top Dispensers
The continuous bottle-top burette enables continuous titration, which leads to rapid, convenient, and accurate results.
  • Long operating life from two easily-replaced micro-batteries 1,5 V
  • Large, easy-to-read digits
  • Simple, media-specific readjustment via keypad
  • Continuous and precise regulation of titration speed with a large practical thumbwheel
  • Reduced titrant waste with innovative recirculation valve
  • Adaptable to bottles of different sizes
  • Telescopic discharge tube which can be adjusted in both height and length
  • Telescopic, extractable intake tube adapts