Single channel pipettes, mechanical, fixed / variable volume, Research® plus (General Lab Product)
Supplier: EPPENDORF
3124000.016 3124000.024 3124000.032 3124000.040 3124000.059 3124000.067 3124000.075 3124000.083 3124000.091 3124000.105 3124000.113 3124000.121 3123000.012 3123000.020 3123000.039 3123000.047 3123000.055 3123000.063 3123000.071 3123000.080 3123000.098 3123000.101 3123000.144 3123000.942
EPPE3124000.016EA 195 EUR
EPPE3124000.016 EPPE3124000.024 EPPE3124000.032 EPPE3124000.040 EPPE3124000.059 EPPE3124000.067 EPPE3124000.075 EPPE3124000.083 EPPE3124000.091 EPPE3124000.105 EPPE3124000.113 EPPE3124000.121 613-5893 613-5894 613-5895 613-5897 613-5898 613-5900 613-5901 613-5902 613-5896 613-5899 613-6973 613-7085
Single channel pipettes, mechanical, fixed / variable volume, Research® plus (General Lab Product)
Pipettes Micropipettes
Versatile, ergonomic pipette with innovative design, meeting the highest needs in precision and accuracy.
  • Extremely lightweight
  • Innovative re-designed principle of piston-stroke pipetting, piston made of Fortron® with great chemical, thermal and mechanical resistance
  • Lowest aspirating, dispensing, blow-out and tip ejection forces
  • Spring loaded retractable nose cone to ensure tight seal and prolong nose cone life
  • 4-digit volume display for ease of use
  • Fully autoclavable
Designed and constructed for general laboratory and research applications in LifeMore Product Information