VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
613-1489EA 345 EUR
613-1489 613-7063 613-7064 613-7065 613-7066 613-7067 613-7068 613-7069 613-7078 613-7079 613-7080 613-7081
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Pipettes Micropipettes
VWR® Ultra-High Performance pipettes offer superior accuracy and precision, with a proven ergonomic design which are both lightweight and durable.
  • Very low plunger forces
  • Universal tip capability
  • Automatic locking volume adjustment wheel
  • Levered tip ejection and colour coded for volume identification
  • Easy in-house calibration and maintenance
  • Fully autoclavable