Baumé hydrometers
Supplier: Alla France
0100FG060/15QP 0100FG070/15QP 0300FG060/15QP 0400TB050/15QP 0400TB070/15QP 010.005FRG 0120FG010/15QP 012.020FRG 012.040FRG 012.050FRG 012.060FRG 012.070FRG 0100FG030/15QP 0100FG040/15QP 0100FG050/15QP 0300FG070/15QP 0050FG015/15QP 0050FG025/15QP 0050FG035/15QP 0150FG075/15QP 0160FG015/15QP 0160FG030/15QP 0160FG045/15QP 0050FG045/15QP 0050FG055/15QP 0050FG065/15QP 0050FG075/15QP 0160FG060/15QP 0160FG075/15QP
ALLA0100FG060/15QPEA 46.1 EUR
ALLA0100FG060/15QP ALLA0100FG070/15QP ALLA0300FG060/15QP ALLA0400TB050/15QP ALLA0400TB070/15QP 614-0305 614-0310 614-0320 614-0340 614-0350 614-0360 614-0370 ALLA0100FG030/15QP ALLA0100FG040/15QP ALLA0100FG050/15QP ALLA0300FG070/15QP ALLA0050FG015/15QP ALLA0050FG025/15QP ALLA0050FG035/15QP ALLA0150FG075/15QP ALLA0160FG015/15QP ALLA0160FG030/15QP ALLA0160FG045/15QP ALLA0050FG045/15QP ALLA0050FG055/15QP ALLA0050FG065/15QP ALLA0050FG075/15QP ALLA0160FG060/15QP ALLA0160FG075/15QP
Baumé hydrometers
Hydrometers
Baumé hydrometers calibrated at 15 °C.