Specific gravity cups, calibrated
Supplier: Industrial Physics
VF2099 VF2100 VF2097 VF2098
614-0715EA 373.25 EUR
614-0715 SIMEVF2100 SIMEVF2097 SIMEVF2098
Specific gravity cups, calibrated
Specific Gravity Bottles
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certifikace: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Delivery information: Supplied with a calibration certificate.