VWR®, Frio Temp Precision Thermometers
Supplier: VWR Collection
620-0843EA 124 EUR
620-0843 620-0844 620-0845 620-0848 620-0854 620-0855 620-0856 620-0878 620-0886 620-0902
VWR®, Frio Temp Precision Thermometers
Thermometers Analogue Thermometers
These glass thermometers can be used to take readings in freezers, refrigerators, incubators, ovens and blood banks.
  • Enviro safe non toxic and liquid or red spirit liquid
  • Accurate temperature reading without sample contamination
  • Individual serial number
  • Wide choice
Accurate temperature reading is achieved by using the filled bottle (included), which mimics the sample. Insert the thermometer into the bottle and place near the samples for best results. PFA coating safely contains glass and liquid in case of breakage.
Traceable to NIST/DKD
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health