VWR®, Bi-Metallic Min./Max. Thermometer
Supplier: VWR Collection
620-0920EA 35.9 EUR
620-0920
VWR®, Bi-Metallic Min./Max. Thermometer
Thermometers Analogue Thermometers
This maximum/minimum thermometer displays the current temperature and registers the maximum and minimum temperatures attained since the last reset. It features colour coded dual scales on a white background and reset knob for fast, multiple readings. Can be used in high moisture or wet environments including greenhouses, sample preparation areas and water treatment facilities.
  • Three distinct arrows
  • Free-standing or wall mountable
W×H: 230×200 mm