VWR® Traceable®, Memory Card Thermometers
Supplier: VWR Collection
620-1963EA 434 EUR
620-1963
VWR® Traceable®, Memory Card Thermometers
Thermometers Digital Thermometers
Memory card reader (supplied) plugs into any USB port for an instant report. Text report includes date/time of day and sensor readings.
  • Captures 5,9 million readings
  • Sealed bottle sensor insulated from temperature changes when refrigerator is opened
  • Solution in sealed bottle generally recognised as safe by FDA
  • Minimum/maximum feature displays highest/lowest readings for any time period
  • Alarm (visual and audible) set by the user for high and low values ​​to the nearest 0,1 °C
  • Unique four-line display: Temperature of detachable bottle probe, ambient