VWR® Traceable®, Digital Data Logging Thermometer, Refrigerator and Freezer
Supplier: VWR Collection
620-2426EA 355 EUR
620-2426
VWR® Traceable®, Digital Data Logging Thermometer, Refrigerator and Freezer
Thermometers Digital Thermometers
Data logging thermometer that can be used in 21 CFR 11 environments. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators.
  • Features min./max. and current temperatures, smart alarm, °F/°C switchable
  • Smart alarm features a visual/audible alarm that continues to alarm even if the unit returns to non alarm conditions
  • Capable of storing over 1 million temperature observations and 10 unique alarm events
  • Triple display simultaneously shows min./max. and current temperatures, °F/°C switchable
  • Status indicators:
More Product Information