Stereo microscope, S-10P
Supplier: MOTIC
PS10100001
630-0144EA 106 EUR
630-0144
Stereo microscope, S-10P
Microscopes
S-10P is a user friendly stereo microscope with dioptre adjustment on the left eye tube.
  • Compact stand with black and white stage plates and clips
  • Removable objectives, Greenough optical system
  • Focusing block with slip clutch and safety stop
Offers crisp and clean images.
Delivery information: Supplied with a dust cover.