VWR® VisiScope® TL384 M, Metallurgical Microscope
Supplier: VWR Collection
630-1939EA 2750 EUR
630-1939
VWR® VisiScope® TL384 M, Metallurgical Microscope
Microscopes
Upright metallurgical microscope with IOS W-plan MET objectives for routine and educational laboratory applications. With LED lighting source both for incident and transmitted illumination.
  • Observation modes: Brightfield (incident and transmitted light), simple polarisation (incident light)
  • Efficient and energy saving LED lamp with brightness control
  • Abbe condenser with iris diaphragm and graduated N.A. scale
  • Specimen stage with tempered glass plate
  • Modern, ergonomic stand, adjustable focusing tension and limit stop
Delivery information: SuppliedMore Product Information