USB camera, Moticam® S12
Supplier: MOTIC
1100600101211
630-3104EA 940 EUR
630-3104
USB camera, Moticam® S12
Cameras
High Grade Moticam® S12 sCMOS USB microscopy camera for ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
Maximum details – If it is about resolution for display or printing, Moticam S12 is your choice. A 25 fps* frame rate manages the largest data volume of all Moticam S models. If the tiniest detail is in your focus, Moticam S12 is your solution.
Informace pro objednávání: To be purchased separately – C-mount adapter not included, this needs to be from theMore Product Information