Upright microscopes, biological, Panthera C2
Supplier: MOTIC
1100104601132
1100104601142
1100104601131
1100104601141
1100104601162
1100104601172
630-3452EA
1566
EUR
630-3452
630-3454
630-3451
630-3453
630-3455
630-3456
Upright microscopes, biological, Panthera C2
Microscopes
SPECIFICATIONS
Model | Panthera C2 Binocular 4×, 10×, 40× (without 100×) | Panthera C2 Trinocular 4×, 10×, 40× (without 100×) | Panthera C2 Binocular | Panthera C2 Trinocular | Panthera C2 simple phase | Panthera C2 phase |
Environmentally Preferable | ||||||
Observation method | Brightfield, polarisation, darkfield, phase contrast, epi-fluorescence LED | Brightfield, polarisation, darkfield, phase contrast, epi-fluorescence LED | Brightfield, polarisation, darkfield, phase contrast, epi-fluorescence LED | Brightfield, polarisation, darkfield, phase contrast, epi-fluorescence LED | Brightfield, polarisation, darkfield, phase contrast, epi-fluorescence LED | Brightfield, polarisation, darkfield, phase contrast, epi-fluorescence LED |
Head | Siedentopf, 25° inclined, 360° rotating, 360° swivelling | Siedentopf, 25° inclined, 360° rotating, 360° swivelling (light split fixed 50:50) | Siedentopf, 25° inclined, 360° rotating, 360° swivelling | Siedentopf, 25° inclined, 360° rotating, 360° swivelling (light split fixed 50:50) | Siedentopf, 25° inclined, 360° rotating, 360° swivelling (light split fixed 50:50) | Siedentopf, 25° inclined, 360° rotating, 360° swivelling (light split fixed 50:50) |
Eyepieces | Widefield UC-WF 10×/22 mm with dioptre adjustment (±5 dioptre) on both eyepieces, and rubber eyecups | Widefield UC-WF 10×/22 mm with dioptre adjustment (±5 dioptre) on both eyepieces, and rubber eyecups | Widefield UC-WF 10×/22 mm with dioptre adjustment (±5 dioptre) on both eyepieces, and rubber eyecups | Widefield UC-WF 10×/22 mm with dioptre adjustment (±5 dioptre) on both eyepieces, and rubber eyecups | Widefield UC-WF 10×/22 mm with dioptre adjustment (±5 dioptre) on both eyepieces, and rubber eyecups | Widefield UC-WF 10×/22 mm with dioptre adjustment (±5 dioptre) on both eyepieces, and rubber eyecups |
Nosepiece | Quintuple, reversed, coded | Quintuple, reversed, coded | Quintuple, reversed, coded | Quintuple, reversed, coded | Quintuple, reversed, coded | Quintuple, reversed, coded |
Objectives | Infinity, CCIS® UC plan achromatic (Pb-free) 4×, 10×, 40× (spring) | Infinity, CCIS® UC plan achromatic (Pb-free) 4×, 10×, 40× (spring) | Infinity, CCIS® UC plan achromatic (Pb-free) 4×, 10×, 40× (spring), 100× (spring, oil immersion) | Infinity, CCIS® UC plan achromatic (Pb-free) 4×, 10×, 40× (spring), 100× (spring, oil immersion) | Infinity, CCIS® UC plan achromatic (Pb-free) 4×, 10×, Ph 40× / 0.65 / S | Infinity, CCIS® UC plan achromatic (Pb-free) 4×, 10×, Ph 40× / 0.65 / S |
Stage | Compact mechanical stage 185×145 mm with built-in low position rackless coaxial stage control | Compact mechanical stage 185×145 mm with built-in low position rackless coaxial stage control | Compact mechanical stage 185×145 mm with built-in low position rackless coaxial stage control | Compact mechanical stage 185×145 mm with built-in low position rackless coaxial stage control | Compact mechanical stage 185×145 mm with built-in low position rackless coaxial stage control | Compact mechanical stage 185×145 mm with built-in low position rackless coaxial stage control |
Condenser | Focusable and centreable Abbe condenser N.A, 0,90/1,25 with slot for contrast sliders | Focusable and centreable Abbe condenser N.A, 0,90/1,25 with slot for contrast sliders | Focusable and centreable Abbe condenser N.A, 0,90/1,25 with slot for contrast sliders | Focusable and centreable Abbe condenser N.A, 0,90/1,25 with slot for contrast sliders | Focusable and centreable Abbe condenser N.A. 0.90/1.25 with slot for contrast sliders, slider PH2 | Phase contrast 5 position (BF, DF, Ph1, Ph2, Ph3) turret condenser |
Focusing system | Coaxial coarse and fine knobs, fine focus precision 2 μm | Coaxial coarse and fine knobs, fine focus precision 2 μm | Coaxial coarse and fine knobs, fine focus precision 2 μm | Coaxial coarse and fine knobs, fine focus precision 2 μm | Coaxial coarse and fine knobs, fine focus precision 2 μm | Coaxial coarse and fine knobs, fine focus precision 2 μm |
Illumination | Koehler LED 3 W and Quartz halogen 6 V/30 W with intensity control; Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator | Koehler LED 3 W and Quartz halogen 6 V/30 W with intensity control; Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator | Koehler LED 3 W and Quartz halogen 6 V/30 W with intensity control; Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator | Koehler LED 3 W and Quartz halogen 6 V/30 W with intensity control; Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator | Koehler LED 3 W and Quartz halogen 6 V/30 W with intensity control; Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator | Koehler LED 3 W and Quartz halogen 6 V/30 W with intensity control; Motic LightTracer: Light memory, sleep mode (auto on-off), nosepiece LED light intensity indicator |
Electrical | 110 - 240 V (CE) | 110 - 240 V (CE) | 110 - 240 V (CE) | 110 - 240 V (CE) | 110 - 240 V (CE) | 110 - 240 V (CE) |
W×D×H | 282×210×394 mm | 282×210×396 mm | 282×210×394 mm | 282×210×396 mm | 282×210×396 mm | 282×210×396 mm |
Weight | 8,4 kg | 8,4 kg | 8,4 kg | 8,4 kg | 8,4 kg | 8,4 kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...