Digital Fluorescence Inverted Microscope for Advanced Cell Culture Experiments, MATEO FL
Supplier: LEICA MICROSYSTEMS
11504171 11504164 11504169 11533332 12730557 11526245 11526246 11533257 11526246_P
LEIM11504171EA 1930 EUR
LEIM11504171 LEIM11504164 LEIM11504169 LEIM11533332 630-3532 LEIM11526245 630-3567 LEIM11533257 630-3566
Digital Fluorescence Inverted Microscope for Advanced Cell Culture Experiments, MATEO FL
Microscopes
Reliable, reproducible, advanced cell checks. Make your cell culture experiments more reliable and consistent with AI-powered algorithms and analysis.
  • Save time and effort with multi-modal fluorescence and transmitted light imaging in a single platform
  • Eliminate guesswork with AI-assisted confluency measurements and increase confidence in your downstream experiments
  • Experience secure data tracking and seamless data transfer
  • Get an accurate, automated cell count that takes just 5 seconds with Mateo FL, saving you an average of 15 minutes of manual cell