Counting chambers, Nageotte
Supplier: GLASWARENFABRIK KARL HECHT
40446001 40446702
630-2741EA 30.9 EUR
630-2741 631-0928
Counting chambers, Nageotte
Slides Counting Chambers
These counting chambers are used for special applications in lumbar fluid.
  • Subdivision: 40 rectangles of 10×0,25 mm each
  • Centre of ruling marked by double lines