Counting chambers, Neubauer
Supplier: LABOR OPTIK
1300000 1310000 1320000 1330000
631-1131EA 111 EUR
631-1131 631-1114 631-1115 631-1116
Counting chambers, Neubauer
Slides Counting Chambers
Unlike the Neubauer improved counting chambers, the counting area in the group squares is limited, by the outermost of the three border lines.
  • Four large squares in the corners are divided into 16 squares each - Leucocyte counting
  • Large middle square divided into 16 group squares
  • Each large square further divided in 16 squares
Counting grid: 9
Large squares: 1 mm² each
Large squares: 1 mm² each