Staining jars acc. to Hellendahl
Supplier: KARTELL
355
631-9130EA 36.3 EUR
631-9130
Staining jars acc. to Hellendahl
Jars Staining Jars
PMP (TPX), transparent.
  • Extension and cover
Staining jar similar to Hellendahl design for 16 standard microscope slides 76×26 mm (in pairs, back to back).