Magnesium sulphate heptahydrate USP, Ph. Eur., BP

Supplier: VWR Chemicals

Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

6527.1000EA 57.3 EUR
6527.1000 6527.5000 6527.9025
Magnesium sulphate heptahydrate USP, Ph. Eur., BP
Magnesium sulphate heptahydrate

Appearance: White or almost white, crystalline powder or brilliant, colourless crystals.


Upozornení: Not for use as Active Pharmaceutical Ingredient (API)

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) ≤ 10 ppm
Residual solvents Passes test
Residual metal catalysts (skip lot): .
As (Arsenic) ≤ 2 ppm
Cd (Cadmium) ≤ 0.1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Li Max. 0.5 ppm
Ni (Nickel) ≤ 6 ppm
Pb (Lead) ≤ 2 ppm
V (Vanadium) ≤ 10 ppm
Conforms to Ph.Eur./USP/BP Passes test
Ph.Eur./BP specifications: .
Assay (dried substance) [Ph.Eur./BP] 99.0 - 100.5 %
Identification A Passes test USP
Identification B Passes test USP
Appearance of solution Passes test Ph.Eur./BP
Acidity or alkalinity Passes test Ph.Eur./BP
Cl (Chloride) [Ph.Eur./BP] ≤ 300 ppm
Fe (Iron) [Ph.Eur./BP] ≤ 20 ppm
Loss on drying (400°C) [Ph.Eur./BP] 48.0 - 52.0 %
USP specifications: .
Assay (by ignition) [USP] 98.0 - 102.0 %
Cl (Chloride) [USP] ≤ 0.014 %
Fe (Iron) [USP] ≤ 20 μg/g
Se (Selenium) [USP] ≤ 30 μg/g
pH (25°C; 50 mg/ml) [USP] 5.0 - 9.2
Loss on ignition (450 ± 25°C) [USP] 40.0 - 52.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR