pH indicator test strips, PEHANON®
Supplier: MACHEREY-NAGEL
90401 90411 90412 90413 90414 90415 90416 90417 90419 90420 90421 90422 90423 90424 904930
662-0214EA 20.4 EUR
662-0214 662-2329 662-0218 662-2330 662-2331 662-0215 662-0216 662-0217 662-2332 662-2333 662-2334 662-2335 662-2336 662-0117 MANA904930
pH indicator test strips, PEHANON®
Test Papers pH Indicator Papers and Test Strips
PEHANON® are special pH test strips that unify the pH indicator and the reference colours on one strip. Any sample colour has the same effect on both the reference colours and the reactive pad. Users can be sure to get accurate readings.
  • Sample colour has no effect on the results
  • No accessories needed - always ready to use
  • Highly economic - colour scale on every single strip
  • Safe usage due to hydrophobic barrier
An invisible hydrophobic barrier above the top colour field prevents capillary action of the test solution, so the handle will remain dry andMore Product Information