Dissolved oxygen probe, StirrOx® G
Supplier: WTW
201425
664-0510EA 1360 EUR
664-0510
Dissolved oxygen probe, StirrOx® G
Electrodes Dissolved Oxygen Electrodes
Self-stirring oxygen probe for simultaneous stirring and measurement.
  • Single-handed operation for rapid series measurements
  • Constant approach flow for high reproducibility
  • Extremely low self-consumption of oxygen
  • Temperature compensation
  • Membrane leakage monitoring
Informace pro objednávání: Inclusive calibration vessel OxiCal®-ST.