Portable water samplers, TP 4 C
Supplier: MAXX
2039024
701-0114EA 3920 EUR
701-0114
Portable water samplers, TP 4 C
Samplers Water Samplers
A portable, battery operated, and compact automated water sampler with a diaphragm pump vacuum collection system. The standard version has an integrated insulated container and distributor for 24×1 L PE bottles and the sample collection is achieved with programmable microprocessor control.
  • Easy to transport with carrying handles
  • Battery provides independence from mains power
  • Sample programming can be time-, flow-, or event-related
  • Manual sample extraction without interruption of programme
  • ~2000 sample extractions per battery charge (20 °C, 1,5 m height, 1