Water testing kit, MINIKIT
Supplier: Lovibond
414100 414130 414140 414150 414180 414230 414240 414250 414260 414310 414320 414340 414160 414270 414170 414350 414360 414110 414220
LOVI414100EA 58.4 EUR
LOVI414100 705-0410 705-0411 705-0413 705-0415 LOVI414230 LOVI414240 LOVI414250 LOVI414260 LOVI414310 LOVI414320 LOVI414340 705-0414 705-0417 705-0420 705-0421 705-0422 LOVI414110 LOVI414220
Water testing kit, MINIKIT
Environmental Test Kits
MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.
  • MINIKITS are based on titrimetric methods
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years
Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.
Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.