RESPIROMETRIC sensor systems for BOD, aerobic and anaerobic respiration
Supplier: VELP SCIENTIFIC
SA10200146 SB10200146 SB10200156 SA10200156 SA102A0166 SA102B0176 SB102A0166 SB102B0176
705-1171EA 2830 EUR
705-1171 705-1172 VELPSB10200156 705-1190 705-1191 705-1192 VELPSB102A0166 VELPSB102B0176
RESPIROMETRIC sensor systems for BOD, aerobic and anaerobic respiration
BOD Measurement Systems
The VELP RESPIROMETRIC sensor is designed to provide a smart, cloud-enabled solution to laboratories to perform multiple aerobic and anaerobic respiration tests. The RESPIROMETRIC sensor has a unique wireless control that transmits data directly to the PC through the wireless DataBox and RESPIROSoft™ software. The RESPIROMETRIC sensor has premium connectivity to the exclusive VELP Ermes Cloud Platform for remote control and remote assistance.
  • Powerful pressure sensor reading results in the range of 500 to 2000 mbar (hPa)
  • Bright 4-digit LED display ensures