Test kits, photometric, Surfactants, cationic, LCK cuvette tests
Supplier: Hach
LCK331
705-3069EA 176 EUR
705-3069
Test kits, photometric, Surfactants, cationic, LCK cuvette tests
Environmental Test Kits
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C