Colony counters
Supplier: IUL Instruments SA
604 90008701 90008700
710-0108EA 980 EUR
710-0108 710-0106 IULA90008700
Colony counters
Colony Counters
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.