Real-Time PCR kit for detection of Vibrio alginolyticus
Supplier: Q BIOANALYTIC
QB-RTI-11-1000 QBRTI-11-10SMP QB-RTI-11-250 QB-RTI-11-50
710-1749EA 0 EUR
710-1749 710-1750 710-1751 710-1752
Real-Time PCR kit for detection of Vibrio alginolyticus
Microbiology Test Systems PCR Tests
Vibrio alginolyticus causes eye, ear and wound infections. It can be tested using this Real-Time PCR test. The analysis can be done in water and food samples. You can use this test in combination with an appropriate pre- enrichment in alkaline peptone-water or you can confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certifikace: DIN EN ISO 20838 conform detection in food, feed and water samples from pre-enrichment cultures or for confirmation of bacterial colonies.