Petri dish racks, Nalgene®
Supplier: Thermo Fisher Scientific
5921-0060
710-4106EA 168 EUR
710-4106
Petri dish racks, Nalgene®
Racks Petri Dish Racks
Petri dish rack with PC frame and white PC posts.
  • Smooth, rounded corners
  • Finger grip handle and rubber feet make carrying safe and secure
  • Rack can be decontaminated by heat sterilisation
For use up to 72 plastic round dishes (60×15 mm). Simple assembly required.
W×D×H: 203×298×152 mm