Staining jars acc. to Hellendahl, WHEATON®
Supplier: DWK Life Sciences
900620
720-0710EA 442 EUR
720-0710
Staining jars acc. to Hellendahl, WHEATON®
Jars Staining Jars
Soda-lime glass.
  • Easy to use due to wide opening
  • For staining or use as a TLC developing chamber
Staining jar holds eight individual 76×26 mm standard slides or 16 slides back to back.