Cell counter cup, Coulter®
Supplier: KARTELL
936
720-0812EA 68.4 EUR
720-0812
Cell counter cup, Coulter®
Sample Containers Sample Cups for Analysers
Polystyrene cup with polyethylene lid graduated at 10 and 20 ml.
  • Graduated at 10 and 20 ml
Ø×H: 30×56 mm
Balení: Cups and lids are packaged separately.