Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Supplier: Simport Scientific
M509-10T M509-11T M509-12T M509-2T M509-3T M509-4T M509-5T M509-6T M509-7T M509-8T M509-9T
720-1422EA 867 EUR
720-1422 720-1424 720-1426 720-1431 720-1433 720-1435 720-1437 720-1439 720-1441 720-1443 720-1445
Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Embedding Cassettes Tissue Processing Cassettes
To be used with Leica and Sakura inkjet printers.
  • The one-piece disposable plastic cover is pre-installed on each cassette
  • Eliminates the need for reusable steel lids
  • The cover can be opened and closed as often as necessary and will always relock
Moulded from a special high density acetal polymer, they keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage. The cover does not protrude aboveMore Product Information