VWR® Q Path® Microtwin, Cassettes
Supplier: VWR Collection
720-2189EA 187 EUR
720-2189 720-2190 720-2184 720-2186 720-2187 720-2188 720-2191 720-2183 720-2185
VWR® Q Path® Microtwin, Cassettes
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassette with mounted disposable lid, supplied in dispenser. Cassettes with two compartments each with pores measuring precisely 0,09 mm². Packed in three boxes of 500 cassettes.
  • No biopsy pads, biopsy capsules or filter paper needed
  • Surface area of pore: 0,3×0,3 mm
  • Hinge
  • 45° writing angle