Sutures, Monosyn® / Safil® / Supramid and Silkam®
Supplier: B. Braun
C0022067 C0712213 C0762113
720-2587EA 1140 EUR
720-2587 720-2588 BRAUC0762113
Sutures, Monosyn® / Safil® / Supramid and Silkam®
Sutures
Synthetic materials for every suture in veterinary surgery.
  • Monosyn® = Mid-term absorbable monofilament and uncoated suture made of glyconate
  • Safil® = Mid-term braided and coated absorbable synthetic suture made of polyglycolic acid
  • Supramid and Silkam® = Non-absorbable synthetic suture
Order Now
SPECIFICATIONS
Model Monosyn® Supramid Silkam
Colour violet dark dark
Environmentally Preferable
Material Glyconate Polyamide 6/6.6 Virgin silk
Coating Aucun Aucun Cire et silicone
Thread structure Monofile Pseudofile Braided, twisted
Thread strength USP 0; metric 3,5 USP 3/0; metric 2 USP 6/0; metric 0,7
Resorption 60 – 90 days Non-absorbable Non-absorbable
Breaking strength 1 day 100 %
14 days 50 %
21 days 20 %
- -
Indication Gynaecology, paediatric surgery, sub-and intracutan Plastic surgery, skin closure Dental surgery, ophthalmic surgery, neurosurgery