SUPERFLO™ Histology Cassettes, One-Piece Moulded
Supplier: Simport Scientific
M531-2 M531-3 M531-4 M531-5 M531-6 M531-7 M531-8 M531-9 M532-10 M430-10T M430-11T M430-12T M430-2T M431-8T M431-9T M432-10T M432-11T M432-12T M432-2T M532-12 M532-2 M532-3 M532-4 M532-5 M532-6 M430-9T M431-10T M431-11T M431-12T M431-2T M431-3T M431-4T M431-5T M431-6T M430-3T M430-4T M430-5T M430-6T M430-7T M430-8T M431-7T M432-3T M432-4T M432-5T M432-6T M432-7T M432-8T M432-9T M530-10 M530-11 M530-12 M530-2 M530-3 M530-4 M530-5 M530-6 M530-7 M530-8 M530-9 M531-10 M531-11 M531-12 M532-11 M532-7 M532-8 M532-9
SIMPM531-2EA 132 EUR
SIMPM531-2 SIMPM531-3 SIMPM531-4 SIMPM531-5 SIMPM531-6 SIMPM531-7 SIMPM531-8 SIMPM531-9 SIMPM532-10 720-3027 720-3026 720-3025 720-3024 SIMPM431-8T SIMPM431-9T SIMPM432-10T SIMPM432-11T SIMPM432-12T SIMPM432-2T SIMPM532-12 SIMPM532-2 SIMPM532-3 SIMPM532-4 SIMPM532-5 SIMPM532-6 720-3017 SIMPM431-10T SIMPM431-11T SIMPM431-12T SIMPM431-2T SIMPM431-3T SIMPM431-4T SIMPM431-5T SIMPM431-6T 720-3023 720-3022 720-3021 720-3020 720-3019 720-3018 SIMPM431-7T SIMPM432-3T SIMPM432-4T SIMPM432-5T SIMPM432-6T SIMPM432-7T SIMPM432-8T SIMPM432-9T 720-3016 720-3015 720-3014 720-3013 720-3012 720-3011 720-3010 720-3009 720-3008 720-3007 720-3006 SIMPM531-10 SIMPM531-11 SIMPM531-12 SIMPM532-11 SIMPM532-7 SIMPM532-8 SIMPM532-9
SUPERFLO™ Histology Cassettes, One-Piece Moulded
Embedding Cassettes Tissue Processing Cassettes
This model is a one-piece moulded cassette so no separate lids to manage. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss.
  • Unique lateral vents, fluids can access tissue from all 4 sides
  • Basket style for small tissue management
  • One-piece moulded with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colours
Moulded from high-density polymer, it keeps specimens safely submerged in liquid.
Available for tissue, biopsy and micro-biopsy. This cassette model is supplied in bulk and also inMore Product Information