Gel tools
Supplier: UVP ULTRA VIOLET PRODUCTS
85-0005-01 85-0006-01 85-0007-01 85-0002-01 85-0003-01 33-0014-02
730-1021EA 97.9 EUR
730-1021 730-1022 730-1023 700-1440 700-1444 733-2011
Gel tools
Tool Sets
Gel cutter: The gel cutter's edge allows for easy cutting and removal of gel material.
Gel ruler: The gel ruler has centimetre markings that fluoresce under 365 and 302 nm UV wavelengths, providing reference marks for DNA analysis.
Gel scooper: Made of strong acrylic with a bevelled edge, the gel scooper is designed for easy transfer of gels from electrophoresis equipment to viewing equipment.
Gel trays: Gel trays (in two sizes), made of UV transmitting Plexiglass, can be used for moving gels to the transilluminator. The tray protects the transilluminator filter surface fromMore Product Information