ALPS™ 3000 Automated Microplate Heat Sealer
Supplier: THERMO MATRIX TECHNOLOGIES
AB-3000
731-0201EA 35456 EUR
731-0201
ALPS™ 3000 Automated Microplate Heat Sealer
Heat Sealers Microplate Heat Sealers
The ALPS™ 3000 microplate heat sealer is a fast, compact heat sealer created for optimal robotic integration in high throughput environments, whilst allowing manual, benchtop control.
  • Seals are top-loaded and easily accessible, with rapid plate sealing action
  • Touch screen technology for easy operation and clear fault diagnosis
  • Can be integrated with the majority of commercially available plate stackers
Equipped with both on-board touch screen operation for bench top use and an RS232 port, plus programming parameters for robotic integration. SensorsMore Product Information