Optical bottom plates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
142761 142762 242763 242764 265301 265302 240074 164586 265300
734-2031EA 686 EUR
734-2031 734-2032 734-2180 734-2181 732-2741 732-2742 734-2178 734-2086 732-2740
Optical bottom plates, 96- and 384-well, Nunc™
Microplates HTS Microplates
PS upper structure with either PS film or no. 1.5 coverglass base. Designed for fluorescence and luminescence based assays, these plates have white or black upper structures with optically clear bottoms. Consequently, they can be read from bottom and top, or both.
  • Compatible with automated handling equipment
  • Alphanumeric well identification
  • Optimised for scintillation counting
  • Low fluorescence background ensures accurate results, due to higher signal-to-noise ratios
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement