PCR tubes, 0,1 ml, with or without cap strips, Axygen®
Supplier: Corning
PCR-0104-C PCR-0108-LP-C PCR-0108-LPRTC PCR-0108-LPRTW
732-2422EA 232.6 EUR
732-2422 AXYGPCR-0108-LP-C 732-2913 732-2424
PCR tubes, 0,1 ml, with or without cap strips, Axygen®
Tubes PCR Tubes
Low profile eight strips PCR tubes ideal for low sample volumn reactions. Low profile design allows reduced reaction time and evaporation due to less dead air volume.
  • Low profile design results in faster and more efficient PCR reactions
  • Tubes available in white or clear, with or without cap
  • Leak-Proof Seals
  • Precise thermal transfer
  • Certified DNase-/RNase-free and nonpyrogenic
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa