Peqlab peqGOLD, Deoxynucleotide Triphosphate (Solution)

Supplier: Peqlab brand

732-3160EA 0 EUR
732-3160 732-3161 732-3155 732-3171
Peqlab peqGOLD, Deoxynucleotide Triphosphate (Solution)
Nucleic Acid Reagents Nucleotides

Individually packaged deoxynucleotide triphosphates manufactured to the highest quality levels.


  • In solution and ready to use, pH 7,5
  • Synthetically manufactured
  • Purity, measured by HPLC, >99%
  • Highly stable, >90% stability after 6 weeks storage at room temperature


Certifikace: Manufacturing and quality control under ISO 9001 and ISO 14001.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR