Strips of 8 PCR tubes with attached flat individual caps
Supplier: Brand
781316 781317 781315 781318
732-1504EA 161 EUR
732-1504 732-1505 732-3613 732-3614
Strips of 8 PCR tubes with attached flat individual caps
Tubes PCR Tubes
PP. Eight connected 0,2 or 0,15 ml PCR tubes with attached flat individual caps. The tubes are held together by three connectors for increased rigidity.
  • Cap offers protection against contamination
  • Highly transparent cap makes them ideal for real-time PCR (qPCR)
  • Available in standard or low profile versions
  • Free from DNase, RNase and DNA
Colour: Clear or white