Hybridisation bottles, WHEATON®
Supplier: DWK Life Sciences
805000 805001 805003 805007 805011 805015 805021 805027 805031
732-4600EA 123 EUR
732-4600 732-4601 732-4602 732-4603 732-4604 732-4605 732-4607 732-4609 732-4610
Hybridisation bottles, WHEATON®
Bottles Hybridisation Bottles/Tubes
Borosilicate glass bottles, PBT caps with PTFE/silicone liners.
  • Five bottle sizes to accommodate a variety of membrane sizes
  • Bottles fit most standard hybridisation ovens
  • Heavy 3,2 mm side walls provide added safety
  • Safety-coated bottles are available with a strong, slip-resistant plastic coating to reduce potential breakage and exposure to hazardous materials
  • Acrylic rack for holding up to six hybridisation bottles available for safe transport or laboratory storage
Useful in nucleic acid and protein blotting procedures that require a minimum of probeMore Product Information