Microscope adapters for micromanipulators
Supplier: EPPENDORF
5181212.006 5181201.004 5181220.009 5181202.000 5181233.003 5181237.009
631-0217EA 0 EUR
631-0217 631-0213 631-0218 631-0214 631-0224 732-6016
Microscope adapters for micromanipulators
Adapters Adapters, Camera to Microscope
Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microscopes, independent of the microscope tripod used.