ABsolute Blue™ qPCR Mixes

Supplier: Thermo Fisher Scientific

AB-4136/B AB-4137/A AB-4138/B AB-4139/A AB-4318/B AB-4319/A AB-4322/B AB-4323/A AB-4162/B AB-4163/A AB-4166/B AB-4167/A
733-0943EA 2270 EUR
733-0943 733-0944 733-0949 733-0950 733-0973 733-0974 733-0979 733-0980 733-0955 733-0956 733-0961 733-0962
ABsolute Blue™ qPCR Mixes
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits

ABsolute Blue™ Master Mix incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy and foolproof. Applications include gene expression, siRNA validation, SNP typing and genotyping.


  • Specially formulated to achieve ultimate sensitivity and reproducible qPCR data across all qPCR platforms
  • All mixes contain Thermo-Start® hot start Taq polymerase for increased sensitivity, and dTTP for increased reaction efficiency
  • Easy to identify mis-aliquoted or skipped wells, particularly when white opaque plates or tubes are used


Storage: Store below 0 °C, preferably at −20 °C, in a constant temperature freezer.

Order Now

SPECIFICATIONS

Application qPCR dye
Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR